Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572724
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572724 Supplier Invitrogen™ Supplier No. PA572724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SPATA4
Gene Accession No. Q8K3V1, Q8NEY3
Gene Alias 1700001N01Rik; Spata4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2
Gene Symbols SPATA4
Host Species Rabbit
Immunogen SPATA4 antibody for an 18 amino acid synthetic peptide near the carboxy terminus of human SPATA4. The immunogen is within amino acids 200 - 250 of SPATA4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132851, 69281
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.