Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596732
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596732 Supplier Invitrogen™ Supplier No. PA596732
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAGQEKGY LTQTAAALDK SPSLSPQLAA PIRGRPKKCL VYPHAPKSSR LSRSVLRWLQ GLDLSFFPRN INRDFSNGFL IAEIFCIYYP WELELSSFEN GTSLKVKLDN WAQLEKFLAR KKFKLPKELI HGTIHCKAGV PEILIEEVYT LLTHREIKSI QDDFVNFTDY SYQMRLPLVS RSTVSKSIKD NIRLSELLSN PNMLTNELKA EFLILLHMLQ RKLGRKLNPE WFDVKPTVGE VTLNHLPAQA; Positive Samples: Mouse heart, Rat heart.

The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPATA4
Gene Accession No. Q8K3V1, Q8NEY3
Gene Alias 1700001N01Rik; Spata4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2
Gene Symbols SPATA4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human SPATA4 (NP_6532452).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132851, 306441, 69281
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.