Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA5L1 Monoclonal Antibody (16A2)
GREENER_CHOICE

Catalog No. PIMA534952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534952 Supplier Invitrogen™ Supplier No. MA534952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: K562 cell lysates, HCT116, JAR, MG-63, rat seminal vesicle tissue, human skin tissue, human breast carcinoma tissue, human placenta tissue, human small intestine tissue.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA5L1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 16A2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene SPATA5L1
Gene Accession No. Q9BVQ7
Gene Alias AV141009; C130039A10Rik; SPATA5L1; spermatogenesis associated 5 like 1; spermatogenesis associated 5-like 1; spermatogenesis-associated protein 5-like protein 1
Gene Symbols SPATA5L1
Host Species Mouse
Immunogen Recombinant protein within Human SPATA5L1 aa 379-580.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79029
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.