Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA5L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114318 Supplier Invitrogen™ Supplier No. PA5114318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA5L1 Polyclonal Antibody for IHC (P), Western Blot

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA5L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene SPATA5L1
Gene Accession No. Q9BVQ7
Gene Alias AV141009; C130039A10Rik; SPATA5L1; spermatogenesis associated 5 like 1; spermatogenesis associated 5-like 1; spermatogenesis-associated protein 5-like protein 1
Gene Symbols SPATA5L1
Host Species Rabbit
Immunogen Recombinant protein within Human SPATA5L1 aa 379-580.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79029
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.