Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATA6L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558847
Description
Immunogen sequence: FATYQHSTSP GPLDQPLLRE RFHPGSQSTW KNIHERVCSL LTSHRAQLHQ NKEDSTSEVN YIIERPSYPL KKYSLHE Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 56%.

Specifications
SPATA6L | |
Polyclonal | |
Unconjugated | |
SPATA6L | |
bA6J24.2; C9orf68; SPATA6L; spermatogenesis associated 6 like; spermatogenesis associated 6-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4H0 | |
SPATA6L | |
Recombinant protein corresponding to Human SPATA6L. Recombinant protein control fragment (Product #RP-92897). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction