Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583403 Supplier Invitrogen™ Supplier No. PA583403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRIEAETQTE LSFKSELGTA ETKNMTDSEM NIKQASNCVT YDAKEKIAPL PLEGHDSTWD EIKDDALQHS SPRA Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 59%.

The function of the SPATA7 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATA7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA7
Gene Accession No. Q9P0W8
Gene Alias AI661438; B230306G18Rik; epididymis secretory protein Li 296; fertility related protein WMP1; fertility-related protein WMP1; HEL-S-296; HSD3; HSD-3.1; LCA3; RSD-3; si:ch211-51o12.1; SPATA7; sperm DNA no.3; spermatogenesis associated 7; spermatogenesis-associated protein 7; Spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7 homolog; spermatogenesis-associated protein 7; spermatogenesis-associated protein HSD3; Wmp1
Gene Symbols SPATA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPATA7. Recombinant protein control fragment (Product #RP-95555).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.