Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989842
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9842 Supplier Bioss Supplier No. BS17636R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen SPATC1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene SPATC1
Gene Accession No. Q76KD6
Gene Alias 1700084J23Rik; SPATA15; Spatc1; Speriolin; spermatogenesis and centriole associated 1; Spermatogenesis and centriole-associated protein 1; Spermatogenesis-associated protein 15; Spermatogenic cell-specific Cdc20-binding protein; SPRN; testicular tissue protein Li 181
Gene Symbols SPATC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPATC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375686
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.