Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556128 Supplier Invitrogen™ Supplier No. PA556128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NADLKKQVRL LKENQMLRRL LSQSCQEGGG HDLLPPRAHA YPEAGSPGSG VPDFGRFTSV ADTPSQLQTS SLEDLLCSHA PLSSEDD Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

The function of this protein has not been specifically defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATC1L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATC1L
Gene Accession No. Q9H0A9
Gene Alias 1700022B01Rik; 1700027D21Rik; AA062323; C1H21orf56; C21orf56; C31H21orf56; hypothetical protein LOC522100; Spatc1l; Speriolin-like protein; spermatogenesis and centriole associated 1 like; spermatogenesis and centriole associated 1-like; spermatogenesis and centriole-associated protein 1-like protein; uncharacterized protein C21orf56 homolog
Gene Symbols SPATC1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPATC1L. Recombinant protein control fragment (Product #RP-95895).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84221
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.