Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546062 Supplier Invitrogen™ Supplier No. PA546062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: STTCGRQLEK VPEKRDSGMT EVERTYSANC SDFLESKGCF ANTTPSGKSV Sequence homology: Dog: 85%; Horse: 85%; Human: 100%.

mRNA is expressed during spermatogenesis with highest expression in first meiotic prophase; contains a serine stretch and may be phosphorylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Spats1
Gene Accession No. Q496A3
Gene Alias 1700011H05Rik; 4933400B06Rik; Daip2; DDIP; Dvl2-DEP domain interacting protein; dvl2-DEP domain-interacting protein; SPATA8; SPATS1; spermatogenesis associated serine rich 1; spermatogenesis associated, serine-rich 1; spermatogenesis-associated serine-rich protein 1; SRSP1
Gene Symbols Spats1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SPATS1 (aa 21-70).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221409
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.