Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPATS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562032
Description
Immunogen sequence: SPSMLTGNSP RGCRLPSISS TTCGRQLEKV PEKRDSGMTE VERTYSANCS DFLESKGCFA NTTPSGKSVS SSSS Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.
mRNA is expressed during spermatogenesis with highest expression in first meiotic prophase; contains a serine stretch and may be phosphorylated.
Specifications
SPATS1 | |
Polyclonal | |
Unconjugated | |
Spats1 | |
1700011H05Rik; 4933400B06Rik; Daip2; DDIP; Dvl2-DEP domain interacting protein; dvl2-DEP domain-interacting protein; SPATA8; SPATS1; spermatogenesis associated serine rich 1; spermatogenesis associated, serine-rich 1; spermatogenesis-associated serine-rich protein 1; SRSP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q496A3 | |
Spats1 | |
Recombinant protein corresponding to Human SPATS1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction