Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110419 Supplier Invitrogen™ Supplier No. PA5110419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRKQNQKDS SGFIFDLQSN TVLAQGGAFE NMKEKINAVR AIVPNKSNNE IILVLQHFDN CVDKTVQAFM EGSASEVLKE WTVTGKKKNK KKKNKPKPAA EPSNGIPDSS KSVSIQEEQS APSSEKGGMN GYHVNGAIND TESVDSLSEG LETLSIDARE LEDPESAMLD TLDRTGSMLQ NGVSDFETKS LTMHSIHNSQ QPRNAAKSLS RPTTETQFSN MGMEDVPLAT SKKLSSNIEK.

SPATS2 (Spermatogenesis Associated Serine Rich 2) is a Protein Coding gene. Diseases associated with SPATS2 include Progesterone-Receptor Negative Breast Cancer. An important paralog of this gene is SPATS2L. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SPATS2
Gene Accession No. 0
Gene Alias Nbla00526; P59SCR; putative protein product of Nbla00526; SCR59; Serine-rich spermatocytes and round spermatid 59 kDa protein; serine-rich spermatocytes and round spermatid protein; SPATA10; SPATS2; spermatogenesis associated serine rich 2; spermatogenesis associated, serine rich 2; spermatogenesis associated, serine-rich 2; spermatogenesis-associated serine-rich protein 2
Gene Symbols SPATS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SPATS2 (NP_075559.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300221
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.