Learn More
SPATS2/SPATA10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304970AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene.Specifications
SPATS2/SPATA10 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q86XZ4 | |
SPATS2 | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SPATS2 | |
Spermatogenesis-associated serine-rich protein 2, Serine-rich spermatocytes and round spermatid 59 kDa protein, p59scr, SPATS2, SCR59, SPATA10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65244 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.