Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPATS2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566836 Supplier Invitrogen™ Supplier No. PA566836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEITHPKNNYS SRTPCSSLLP LLNAHAATSG KQSNFSRKSS THNKPSEGKA ANPKMVSSLP STADPSHQTM PANKQNGSSN QRRRFN Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.

SPATS2L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPATS2L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATS2L
Gene Accession No. Q9NUQ6
Gene Alias 2810022L02Rik; A230104H11Rik; AA987173; AI480531; AI842453; AW413586; DNA polymerase transactivated protein 6; DNA polymerase-transactivated protein 6; DNAPTP6; RGD1309930; SGNP; SP1224; Spats2l; SPATS2-like protein; spermatogenesis associated serine rich 2 like; spermatogenesis associated, serine rich 2 like; spermatogenesis associated, serine-rich 2-like; stress granule and nucleolar protein
Gene Symbols SPATS2L
Host Species Rabbit
Immunogen Recombinant Human SPATS2L. Recombinant protein control fragment (Product #RP-105390).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.