Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATS2L Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569588
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-9588 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9588 Supplier Bethyl Laboratories, Inc Supplier No. A305118AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

SPATS2L is a protein coding gene.

Specifications

Antigen SPATS2L
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SPATS2L
Gene Accession No. Q9NUQ6
Gene Alias DNA polymerase transactivated protein 6, DNA polymerase-transactivated protein 6, DNAPTP6, SGNP, spermatogenesis associated, serine rich 2 like, spermatogenesis associated, serine-rich 2-like, stress granule and nucleolar protein
Gene Symbols SPATS2L
Host Species Rabbit
Immunogen Between 508 and 558
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26010
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.