Learn More
SPATS2L Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305152A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SPATS2L is a protein coding gene.Specifications
SPATS2L | |
Polyclonal | |
Unconjugated | |
SPATS2L | |
DNA polymerase transactivated protein 6, DNA polymerase-transactivated protein 6, DNAPTP6, SGNP, spermatogenesis associated, serine rich 2 like, spermatogenesis associated, serine-rich 2-like, stress granule and nucleolar protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26010 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9NUQ6 | |
SPATS2L | |
Between 450 and 500 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.