Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATS2L Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569674
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9674 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9674 Supplier Bethyl Laboratories, Inc Supplier No. A305152AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SPATS2L is a protein coding gene.

Specifications

Antigen SPATS2L
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene SPATS2L
Gene Accession No. Q9NUQ6
Gene Alias DNA polymerase transactivated protein 6, DNA polymerase-transactivated protein 6, DNAPTP6, SGNP, spermatogenesis associated, serine rich 2 like, spermatogenesis associated, serine-rich 2-like, stress granule and nucleolar protein
Gene Symbols SPATS2L
Host Species Rabbit
Immunogen Between 450 and 500
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26010
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.