Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Spatula, 6 Pk.
SCI_ED

Catalog No. S2335016 Shop All Fischer Technical Company Products
Change view
Click to view available options
Color:
Red
Transparent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S2335016 Transparent
S2335017 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S2335016 Supplier Fischer Technical Company™ Supplier No. SILSPATSPK6

For use with a stirring rod to help spread or scoop chemicals safely.

Attaches to a stirring rod and is used as a soft style spatula with excellent chemical resistance.

  • Available In both natural rubber and chemically resistant silicone
  • Opening is 5 mm for a stirring rod

Specifications

Quantity 6 Pk.
Autoclavable Not Autoclavable
Shape Square
Color Transparent
Product Type Spatula
Length (Metric) Blade 3.5 cm
Blade Material Silicone
Material Silicone
Length (Metric) 31.75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.