Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Spatula, Stainless Steel, Flat And Spoon Ends
SDPSCI_ED

Catalog No. S16029 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
4 in.
6 in.
8 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S16029 8 in.
S15878 4 in.
S15820 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S16029 Supplier United Scientific™ Supplier No. SSSP08
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical option for everyday lab use.

  • Stainless steel spatulas.
  • One flat end and one spoon end.
  • Available in three sizes

Specifications

Product Type Spatula
Length (English) 8 in.
Material Stainless Steel
Length (Metric) 20.3 cm
Sterility Non-sterile
Autoclavable Autoclavable
Shape Flat Spatula End, Spoon End
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.