Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC™ Absorbents SpillFix Granular

Catalog No. p-9034009 Shop All Brady Corporation Products
Change view
Click to view available options
Size:
20 lb. bag
3 qt. bottle
7 lb. bag
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-666-506 3 qt. bottle
19-666-505 7 lb. bag
19-666-504 20 lb. bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-666-506 Supplier Brady™ Supplier No. SFJARCS

Brady SpillFix granular is completely natural producing no carcinogens or dust. With more than four times the absorption power of clay granular, this product is perfect for both indoor and outdoor use.

  • Premium absorbent made of 100% recycled material
  • Effective on the absorption of hydrocarbons, water-based fluids and most aggressive chemicals
  • Certified safe for landfill disposal

Specifications

Certifications/Compliance OSHA 29 CFR 1910.22(a)(2) ; EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii)
Capacity (English) 0.75 gal.
Capacity (Metric) 2.84 L
Material Biodegradable Granular
Type Absorbent
Format Granular
For Use With (Application) Absorption of hydrocarbons, water-based fluids and most aggressive chemicals
Size 3 qt. bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.