Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SPC HandySorb™ Mop System and Supplies

Catalog No. p-8793002
Click to view available options
Product Type:
Absorbent Pads
Absorbent Pads, Mop Adapter
Absorbent Pillows
Mop, Absorbent Pads

SPC HandySorb Mop System™ is ideal for cleaning hard-to-reach spills. Eliminates kneeling and gets the job done in three simple steps – snap on, absorb and release.

  • Keep hands out of the mess with the No-Touch Universal Pads and Pillows that clean up a variety of fluids
  • Increase reach – handle extends to 6 feet
  • Patented quick-release operation makes disposal of used pads simple and keeps hands clean.

Specifications

Product Type Absorbent Pads
Color Gray
Sterility Non-sterile
Autoclavable Not Autoclavable
Width (English) 19 in.
Quantity 25
For Use With (Application) HandySorb Mop
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.