Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPC24 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297666PE
Description
SPC24 Polyclonal antibody specifically detects SPC24 in Human samples. It is validated for Western BlotSpecifications
SPC24 | |
Polyclonal | |
Western Blot | |
FLJ90806, hSpc24, kinetochore protein Spc24, SPBC24, SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 24 homolog, spindle pole body component 24 homolog (S. cerevisiae) | |
Produced in rabbits immunized with E. coli-derived Human SPC24 fragment. | |
0.1 mL | |
Apoptosis, Cell Biology, Cell Cycle and Replication | |
147841 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction