Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPC24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112225
Description
SPC24 Polyclonal Antibody for Western Blot
Acts as a component of the essential kinetochore-associated NDC80 complex, which is required for chromosome segregation and spindle checkpoint activity. Required for kinetochore integrity and the organization of stable microtubule binding sites in the outer plate of the kinetochore.
Specifications
SPC24 | |
Polyclonal | |
Unconjugated | |
SPC24 | |
2410030K01Rik; AV109292; hSpc24; kinetochore protein Spc24; SPBC24; SPC24; SPC24, NDC80 kinetochore complex component; SPC24, NDC80 kinetochore complex component, homolog; SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae); spindle pole body component 24 homolog | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
147841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q8NBT2 | |
SPC24 | |
E. coli-derived Human SPC24 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction