Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPC25 Antibody (OTI3D11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274311B
Description
SPC25 Monoclonal specifically detects SPC25 in Human samples. It is validated for Western Blot.Specifications
SPC25 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SPC25 | |
Full length recombinant protein of human SPC25 (NP_065726) produced in E.coli. | |
0.1 mL | |
57405.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3D11 | |
Western Blot | |
AD024, hSpc25, kinetochore protein Spc25, MGC22228,2600017H08Rik, SPBC25, SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 25 homolog, spindle pole body component 25 homolog (S. cerevisiae) | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction