Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPC25 Antibody (OTI3D11), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274311C
Description
SPC25 Monoclonal specifically detects SPC25 in Human samples. It is validated for Western Blot.Specifications
| SPC25 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPC25 | |
| Full length recombinant protein of human SPC25 (NP_065726) produced in E.coli. | |
| 0.1 mL | |
| 57405.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3D11 | |
| Western Blot | |
| AD024, hSpc25, kinetochore protein Spc25, MGC22228,2600017H08Rik, SPBC25, SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 25 homolog, spindle pole body component 25 homolog (S. cerevisiae) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction