Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPC25 Monoclonal Antibody (OTI3D11), TrueMAB™, OriGene
SDP

Catalog No. 501681160
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1160 Supplier OriGene Supplier No. TA806559
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein that may be involved in kinetochore-microtubule interaction and spindle checkpoint activity.

Specifications

Antigen SPC25
Applications Western Blot
Classification Monoclonal
Clone OTI3D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SPC25
Gene Accession No. Q9HBM1
Gene Alias AD024, SPBC25, hSpc25
Gene Symbols SPC25
Host Species Mouse
Immunogen Full length human recombit protein of human SPC25 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57405
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.