Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPC25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561444 Supplier Invitrogen™ Supplier No. PA561444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLGLEIRKIY GEKLQFIFTN IDPKNPESPF MFSLHLNEAR DYEVSDSAPH LEGLAEFQEN VRKTNNFSAF LANVRKAFTA TVY Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 83%.

This gene encodes a protein that may be involved in kinetochore-microtubule interaction and spindle checkpoint activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPC25
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPC25
Gene Accession No. Q9HBM1
Gene Alias 2600017H08Rik; 2610205L13Rik; AD024; hSpc25; kinetochore protein Spc25; RGD1307811; SPBC25; SPC25; SPC25, NDC80 kinetochore complex component; SPC25, NDC80 kinetochore complex component, homolog; SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae); spindle pole body component 25 homolog
Gene Symbols SPC25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPC25. Recombinant protein control fragment (Product #RP-98498).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57405
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.