Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPCS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549486
Description
SPCS1 Polyclonal Antibody for Western Blot
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
Specifications
SPCS1 | |
Polyclonal | |
Unconjugated | |
SPCS1 | |
HSPC033; Microsomal signal peptidase 12 kDa subunit; signal peptidase 12kDa; signal peptidase complex 12 kDa subunit; signal peptidase complex subunit 1; signal peptidase complex subunit 1 homolog; SPase 12 kDa subunit; SPC1; SPC12; SPCS1; YJR010C-A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28972 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y6A9 | |
SPCS1 | |
KLH conjugated synthetic peptide between 66-100 amino acids from the Central region of human SPCS1. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction