Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPCS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561146 Supplier Invitrogen™ Supplier No. PA561146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPWPIYRRHP LKWLPVQESS TDDKKPG Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.

Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPCS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPCS1
Gene Accession No. Q9Y6A9
Gene Alias HSPC033; Microsomal signal peptidase 12 kDa subunit; signal peptidase 12kDa; signal peptidase complex 12 kDa subunit; signal peptidase complex subunit 1; signal peptidase complex subunit 1 homolog; SPase 12 kDa subunit; SPC1; SPC12; SPCS1; YJR010C-A
Gene Symbols SPCS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPCS1. Recombinant protein control fragment (Product #RP-96324).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28972
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.