Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPCS2 Antibody (OTI1E4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274312AF405
Description
SPCS2 Monoclonal specifically detects SPCS2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SPCS2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPCS2 | |
Recombinant protein fragment corresponding to amino acids 2-64 of human SPCS2 (NP_055567) produced in E.coli. | |
0.1 mL | |
9789.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1E4 | |
Western Blot | |
MGC117366, signal peptidase complex subunit 2, signal peptidase complex subunit 2 homolog (S. cerevisiae), SPC25 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction