Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112906
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112906 Supplier Invitrogen™ Supplier No. PA5112906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPCS2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPCS2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPCS2
Gene Accession No. Q15005
Gene Alias 5730406I15Rik; AA408713; KIAA0102; MGC117366; Microsomal signal peptidase 25 kDa subunit; mKIAA0102; Probable signal peptidase complex subunit 2; RGD1311253; signal peptidase 25kDa subunit; signal peptidase complex 25 kDa subunit; signal peptidase complex subunit 2; signal peptidase complex subunit 2 homolog; signal peptidase complex subunit 2 homolog (S. cerevisiae); signal peptidase complex subunit 2-like protein; SPase 25 kDa subunit; Spc25; Spcs2; wu:fa12g08; zgc:110364
Gene Symbols SPCS2
Host Species Rabbit
Immunogen Recombinant Human Signal peptidase complex subunit 2 protein (7-86AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9789
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.