Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPCS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553052
Description
Immunogen sequence: AAAAVQGGRS GGSGGCSGAG GASNCGTGSG RSGLLDKWKI DDKPVKIDKW DGSAVKNSLD DSAK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum. [UniProt].
Specifications
SPCS2 | |
Polyclonal | |
Unconjugated | |
SPCS2 | |
5730406I15Rik; AA408713; KIAA0102; MGC117366; Microsomal signal peptidase 25 kDa subunit; mKIAA0102; Probable signal peptidase complex subunit 2; RGD1311253; signal peptidase 25kDa subunit; signal peptidase complex 25 kDa subunit; signal peptidase complex subunit 2; signal peptidase complex subunit 2 homolog; signal peptidase complex subunit 2 homolog (S. cerevisiae); signal peptidase complex subunit 2-like protein; SPase 25 kDa subunit; Spc25; Spcs2; wu:fa12g08; zgc:110364 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
293142, 66624, 9789 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15005, Q9CYN2 | |
SPCS2 | |
Recombinant protein corresponding to Human SPCS2. Recombinant protein control fragment (Product #RP-90736). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction