Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPCS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112722 Supplier Invitrogen™ Supplier No. PA5112722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPCS3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SPCS3 is a component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPCS3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPCS3
Gene Accession No. P61009, Q6ZWQ7
Gene Alias 1810011E08Rik; microsomal signal peptidase 22/23 kDa subunit; microsomal signal peptidase 23 kDa subunit; PRO3567; Signal peptidase complex subunit 3; signal peptidase complex subunit 3 homolog; signal peptidase complex subunit 3 homolog (S. cerevisiae); SPase 22 kDa subunit; SPase 22/23 kDa subunit; SPC22; SPC22/23; SPC3; Spcs3; UNQ1841/PRO3567; YLR066W
Gene Symbols SPCS3
Host Species Rabbit
Immunogen Recombinant Human Signal peptidase complex subunit 3 protein (33-180AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60559, 76687
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.