Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPDEF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 114671AP150UL

Description
PDEF (prostate-derived Ets factor) is a 37.5- kDa protein that acts as an androgen-independent transcriptional activator for the PSA (prostate specific antigen) promoter. It also interacts directly with the DNA binding domain of androgen receptor and enhances androgen-mediated activation of the PSA promoter. PDEF was shown to have transcript levels 192-fold higher in the peripheral blood of some breast cancer patients in comparison with normal individuals. PDEF mRNA is also frequently over-expressed in human breast tumors as well as in human ovarian tumors. In ovarian tumors, PDEF expression seems to down-modulate maligt potential, and thus provides a rationale to screen for drugs that induce PDEF expression in epithelial ovarian tumors.Specifications
SPDEF | |
Polyclonal | |
Unconjugated | |
SPDEF | |
PDEF, Prostate derived Ets factor, Prostate specific Ets, PSE, RP11 375E1__A.3, SPDEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25803, 30051, 689210 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95238, Q9WTP3 | |
SPDEF | |
SPDEF Fusion Protein Ag2020 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction