Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPDL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550267 Supplier Invitrogen™ Supplier No. PA550267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CCDC99 / SPDL1 protein.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridizations indicate that this lectin is specifically expressed in keratinocytes. It is expressed at all stages of epidermal differentiation (i.e., in basal and suprabasal layers). The protein was found mainly in stratified squamous epithelium. It is moderately repressed by retinoic acid. The antigen localized to basal keratinocytes, although it was also found at lower levels in the suprabasal layers, where it concentrated to areas of cell-to-cell contact. The cellular localization and its striking down-regulation in cultured keratinocytes imply a role in cell-cell and/or cell-matrix interactions necessary for normal growth control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPDL1
Applications Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SPDL1
Gene Accession No. Q923A2, Q96EA4
Gene Alias 1700018I02Rik; 2600001J17Rik; 2810049B11Rik; AA409762; arsenite-related gene 1 protein; Ccdc99; coiled-coil domain containing 99; coiled-coil domain-containing protein 99; coiled-coil domain-containing protein 99 {ECO:0000255; HAMAP-Rule:MF_03041}; hSpindly; protein Spindly; RGD1306908; Rhabdomyosarcoma antigen MU-RMS-40.4A; rrhabdomyosarcoma antigen protein MU-RMS-40.4A; SPDL1; Spindle apparatus coiled-coil domain-containing protein 1; spindle apparatus coiled-coil domain-containing protein 1 {ECO:0000255; spindle apparatus coiled-coil protein 1; spindly homolog
Gene Symbols SPDL1
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human Spindle apparatus coiled-coil protein 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54908, 70385
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.