Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPDYA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110420 Supplier Invitrogen™ Supplier No. PA5110420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRHNQMCCET PPTVTVYVKS GSNRSHQPKK PITLKRPICK DNWQAFEKNT HNNNKSKRPK GPCLVIQRQD MTAFFKLFDD DLIQDFLWMD CCCKIADKYL LAMTFVYFKR AKFTISEHTR INFFIALYLA NTVEEDEEET KYEIFPWALG KNWRKLFPNF LKLRDQLWDR IDYRAIVSRR CCEEVMAIAP THYIWQRERS VHHSGAVRNY NRDEVQLPRG PSATPVDCSL CGKKRRYVRL GLSSSSSLSS HTAGVTEKHS QDSYNSLSMD IIGDPSQAYT GSEVVNDHQS NKGKKTNFLK KDKSMEWFTG SEE.

Speedy A, also known as SPDYA, SPDY1, Ringo3 or SPY1, is a 313 amino acid protein that localizes to the nucleus and belongs to the speedy/ringo family. Expressed at high levels in testis and at lower levels in brain, kidney, heart, bone marrow, colon, lung, liver spleen and placenta, Speedy A functions to regulate the G1/S phase transition of the cell cycle, specifically by binding to and activating Cdc2 p34, Cdk2 and p27. Additionally, Speedy A mediates cell survival during DNA damage, suggesting that Speedy A plays a role in proper cell cycle progression throughout the life of the cell. Multiple isoforms of Speedy A exist due to alternative splicing events. The gene encoding Speedy A maps to human chromosome 2, which encodes over 1,400 genes and comprises nearly 8% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPDYA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPDYA
Gene Accession No. Q5MJ70
Gene Alias 4921517J08Rik; 4930548B21Rik; Gs4; hSpy/Ringo A; Lm23; MLZ-465; mSpy/Ringo A; protein expressed in male leptotene and zygotene spermatocytes 465; Rapid inducer of G2/M progression in oocytes A; RINGO A; RINGO3; RINGOA; SPDY1; SPDYA; spdya {ECO:0000250; speedy A; speedy A1; speedy A2; speedy homolog 1; speedy homolog A; speedy homolog A (Xenopus laevis); speedy protein A; speedy/ringo; speedy/RINGO cell cycle regulator family member A; Speedy-1; spermatogenesis related protein Gs4; spy1; UniProtKB:Q5MJ70}
Gene Symbols SPDYA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-313 of human SPDYA (NP_877433.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245711
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.