Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPDYC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558963 Supplier Invitrogen™ Supplier No. PA558963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPLVTTQVEL GGCSRQGGGN GFLRFRQHQE VQAFLSLLED SFVQEFLSKD PCFQISDKY Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.

Promotes progression through the cell cycle via binding and activation of CDK1 and CDK2. Involved in the spindle-assembly checkpoint. Required for recruitment of MAD2L1, BUBR1 and BUB1 to kinetochores. Required for the correct localization of the active form of Aurora B in prometaphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPDYC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPDYC
Gene Accession No. Q5MJ68
Gene Alias hSpy/Ringo C; rapid inducer of G2/M progression in oocytes C; RINGO C; Ringo2; RINGOC; SPDYC; speedy; speedy C; speedy homolog C; speedy protein C; speedy/RINGO cell cycle regulator family member C
Gene Symbols SPDYC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPDYC. Recombinant protein control fragment (Product #RP-97558).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.