Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPDYE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562470 Supplier Invitrogen™ Supplier No. PA562470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLVLPEHHEA FNRLLEDPVI KRFLAWDKDL RVSDKYLLAM Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.

This gene is located at chromosome 7p13 which is close to the Williams Beuren syndrome chromosome region 7q11.23. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPDYE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPDYE1
Gene Accession No. Q8NFV5
Gene Alias putative WBSCR19-like protein 6; Ringo1; SPDYE; SPDYE1; speedy; Speedy E; speedy homolog E1; speedy protein E1; speedy/RINGO cell cycle regulator family member E1; WBSCR19; Williams Beuren syndrome chromosome region 19 protein; williams-Beuren syndrome chromosomal region 19 protein
Gene Symbols SPDYE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPDYE1. Recombinant protein control fragment (Product #RP-104468).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.