Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPDYE2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571260
Description
This target displays homology in the following species: Human: 100%
SPDYE2B gene ontology annotations related to this gene include protein kinase binding.
Specifications
SPDYE2B | |
Polyclonal | |
Unconjugated | |
SPDYE2B | |
putative speedy protein E2-like protein; putative WBSCR19-like protein 3; SPDYE2B; SPDYE2L; speedy protein E2B; speedy/RINGO cell cycle regulator family member E2B; WBSCR19-like protein 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100310812 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NHP3 | |
SPDYE2B | |
synthetic peptide directed towards the C-terminal region of human SPDYE2B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction