Learn More
MilliporeSigma™ Supelco™ SPE Tube Adapter configured for AutoTrace Automated Systems
Catalog No. 111012129 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
3 mL Tubes
6 mL Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11-101-2129 6 mL Tubes
11-102-0922 3 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2129 Supplier MilliporeSigma™ Supelco™ Supplier No. 57126
Add to Cart
Add to Cart
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 6/Pk.
For Use With 6 mL Tubes