Learn More
SPECC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302412A

Description
The recommended shelf life for this product is 1 year from date of receipt.
NSP type 5alpha/3alpha is a novel structure protein that may play a role in nuclear function during cell division. It is also highly expressed in certain tumor cell lines, suggesting that it may serve as a good tumor marker.Specifications
SPECC1 | |
Polyclonal | |
Unconjugated | |
SPECC1 | |
NSP5a3a, NSP 5 alpha 3 alpha, NSP5alpha3alpha, Specc1, HCMOGT-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92521 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5M775 | |
SPECC1 | |
Between 125 and 175 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.