Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPECC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590714 Supplier Invitrogen™ Supplier No. PA590714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHLRNELRDM RAQLGINEDH SEGDEKSEKE TIMAHQPTDV ESTLLQLQEQ NTAIREELNQ LKNENRMLKD RLNALGFSLE QRLDNSEKLF GYQSLSPEIT PGNQSDGGGT LTSSVEGSAP GSVEDLLSQD ENTLMDHQHS NSMDNLDSEC SEVYQPLTSS DDALDAPSSS ESEGIPSIER SRKGSSGNAS EVSVACLTER IHQMEENQHS TSEELQATLQ ELADLQQITQ ELNSENERLG EEKVILMESL CQQSDKLEHF SRQIEYFRSL LDEHHISYVI DEDVKSGRYM ELEQRYMDLA E; Positive Samples: LO2, HeLa, A375; Cellular Location: Cell junction, Cytoplasm, cytoskeleton, gap junction, spindle.

This gene encodes a protein that is leucine-rich and is thought to play a role in regulating the interaction of the cytoskeleton with a variety of cellular processes. Mutations in this gene are associated with the French-Canadian type of Leigh syndrome. Transcripts ranging in size from 4. 8 to 7. 0 kb which result from alternative polyadenylation have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPECC1L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SPECC1L
Gene Accession No. Q69YQ0
Gene Alias 4930470P14Rik; 4932439K10Rik; 9530057A13Rik; AI317249; cytokinesis and spindle organization A; cytospin A; cytospin-A; CYTSA; GBBB2; Kiaa0376; mKIAA0376; OBLFC1; Renal carcinoma antigen NY-REN-22; RGD1309570; Specc1l; SPECC1-like; SPECC1-like protein; sperm antigen with calponin homology and coiled-coil domains 1 like; sperm antigen with calponin homology and coiled-coil domains 1-like
Gene Symbols SPECC1L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human SPECC1L (NP_0561454).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23384
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.