Learn More
Greiner Bio-One Special Marker Pen
GREENER_CHOICE
Catalog No. p-7075475 Shop All Greiner Bio-One Products
Click to view available options
Color:
Red
Tip Style:
Superfine
Quick-drying ink offered in a variety of colors
  • Fine tip is excellent for general marking
  • Highly resistant to abrasion and water
  • Will not smear when subjected to water or alcohol during normal use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Marker Pen
Tip Style Superfine
Color Red
Material Polyethylene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.