Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Specialty Glass Column: Glass Coarse Frit
Described in US EPA Protocol - Fill columns with appropriate adsorbents when cleaning or drying environmental samples for pesticide or priority pollutant analyses
Supplier: MilliporeSigma™ Supelco™ 64756
Description

Specifications
Specialty Glass Column (SJ Inlet Joint) | |
LPLC | |
1 | |
10.5 mm |
13 mm | |
Glass (Frit), PTFE (Stopcock) | |
Overall Length: 415 mm | |
Frit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction