Learn More
Description
- Available in black
- Available with zipper or drawstring
Specifications
Specifications
| Description | Zippered Storage Case |
| Color | Blue |
| Product Type | Eyewear Case |
| For Use With (Equipment) | For glasses |
| For Use With (Application) | Ideal for lens cleaning |
| Material | Microfiber |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.