Learn More
Spectrum™ Spectra/Chrom™ Tubing Cutter
Description
Cuts to make an even, flat edge
- For all polymer-type tubing (vinyl, fluoroelastomer, silicone, polyethylene, TFE)
- Razor-sharp hardened-steel blade
- Special V socket firmly holds tubing perpendicular to the cutting blade
Specifications
Specifications
| Product Type | Tubing Cutter |
| For Use With (Equipment) | All Polymer-type Plastic tubing |
| Material | Carbon Steel |
| Size | Pocket-Size |
| For Use With (Application) | Ideal for 13mm dia. or smaller tubing and cuts to make an sharp, even, square cut |