Learn More
Thermo Scientific™ Spectra™ VRE Medium
Description
Improve the prevention and control of VRE in healthcare settings by rapidly and reliably isolating and presumptively identifying vancomycin-resistant Enterococcus (VRE) within 24 hours, direct from sample, using Thermo Scientific™ Spectra™ VRE. Vancomycin-resistant Enterococcus faecium and Enterococcus faecalis are differentiated through the utilization of two separate chromogens. Interpretation is aided through the inclusion of a semi-opaque background, providing a distinct visual contrast against light blue and indigo colonies.
Additional antibiotics, in combination with vancomycin, are present to suppress the growth of competing flora including E. gallinarum and E. casseliflavus, both of which are intrinsically resistant to vancomycin, possessing the chromosomally encoded VanC resistance mechanism. The VanC resistance mechanism is not readily transmissible between organisms and, as such, is deemed less clinically significant than VanA and VanB mechanisms which are encoded on freely transmissible genetic elements, plasmids and transposons, thus increasing the risk of resistance genes spreading to other organisms.
- Rapidly and economically detects colonization of Vancomycin-Resistant Enterococcus (VRE) colonization
- Aids in the prevention and controls the spread of vancomycin resistance in healthcare settings
- Correctly classifies patients whether positive or negative
- Provides superior results in a selective and differential chromogenic medium
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Semi-opaque light tan |
| Content And Storage | 2°C to 8°C |
| Format | 85mm Monoplate |
| Manufacturing Origin | United States |
| Media Type | Spectra VRE Medium |
| Product Type | Agar |
| Fill Volume | 18 mL |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.