Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin alpha 1 Antibody (rSPTA1/1832), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308317JF646
Description
Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Spectrin alpha 1 | |
Monoclonal | |
Janelia Fluor 646 | |
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary) (Uniprot: P02549) | |
100 μg | |
Cell Biology, Cellular Markers | |
6708 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
rSPTA1/1832 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction