Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin alpha 1, Mouse anti-Human, Clone: SPTA1/1832, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279772100UG
Description
Spectrin alpha 1 Monoclonal specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Spectrin alpha 1 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot, ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P02549 | |
SPTA1 | |
Recombinant human Spectrin alpha 1 fragment (around aa356-475) (exact sequence is proprietary) | |
100 μg | |
Cell Biology, Cellular Markers | |
6708 | |
Store at 4°C. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
SPTA1/1832 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction