Learn More
Invitrogen™ Spectrin alpha-1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total alpha 1 Spectrin.
Specifications
Specifications
| Antigen | Spectrin alpha-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | SPTA1 |
| Gene Accession No. | P02549, P08032 |
| Gene Alias | 2610027H02Rik; A2a; AF093576; a--fodrin; AI451697; alpha II spectrin; alpha-fodrin; alpha-I spectrin; alpha-II spectrin; alpha-spectrin 1, erythroid; alpha-spectrin 2; alpha-spectrin 2, brain; brain alpha-spectrin; EIEE5; EL2; elliptocytosis 2; erythroid alpha-spectrin; erythroid spectrin alpha; fodrin alpha chain; ha; hemolytic anemia; HPP; HS3; ihj; inhibitory protein factor; IPF; mutant alpha spectrin; NEAS; neuroscience mutagenesis facility, 4; nmf4; noerythroid alpha-spectrin 2; nonerythroid alpha-spectrin 2; spectrin alpha 1; spectrin alpha chain, brain; spectrin alpha chain, erythrocyte; spectrin alpha chain, erythrocytic 1; Spectrin alpha chain, non-erythrocytic 1; spectrin alpha, erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, erythrocytic 1; spectrin, alpha, erythrocytic 1 (elliptocytosis 2); spectrin, alpha, non-erythrocytic 1; spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; sph; SPH3; spherocytosis; Spna1; Spna-1; Spna2; Spna-2; SPTA; Spta1; SPTA2; SPTAN1 |
| Gene Symbols | SPTA1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.