Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin alpha-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144645 Supplier Invitrogen™ Supplier No. PA5144645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: K562 cell lysate, NIH/3T3 cell lysate, A549 cell lysate. Predicted band size: 280 kDa Subcellular Location: Cytoplasm, Cytoskeleton.

Spectrin is an actin crosslinking and molecular scaffold protein that links the plasma membrane to the actin cytoskeleton, and functions in the determination of cell shape, arrangement of transmembrane proteins, and organization of organelles. It is a tetramer made up of alpha-beta dimers linked in a head-to-head arrangement. This gene is one member of a family of alpha-spectrin genes. The encoded protein is primarily composed of 22 spectrin repeats which are involved in dimer formation. It forms weaker tetramer interactions than non-erythrocytic alpha spectrin, which may increase the plasma membrane elasticity and deformability of red blood cells. Mutations in this gene result in a variety of hereditary red blood cell disorders, including elliptocytosis type 2, pyropoikilocytosis, and spherocytic hemolytic anemia. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin alpha-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SPTA1
Gene Accession No. P02549, P08032
Gene Alias 2610027H02Rik; A2a; AF093576; a--fodrin; AI451697; alpha II spectrin; alpha-fodrin; alpha-I spectrin; alpha-II spectrin; alpha-spectrin 1, erythroid; alpha-spectrin 2; alpha-spectrin 2, brain; brain alpha-spectrin; EIEE5; EL2; elliptocytosis 2; erythroid alpha-spectrin; erythroid spectrin alpha; fodrin alpha chain; ha; hemolytic anemia; HPP; HS3; ihj; inhibitory protein factor; IPF; mutant alpha spectrin; NEAS; neuroscience mutagenesis facility, 4; nmf4; noerythroid alpha-spectrin 2; nonerythroid alpha-spectrin 2; spectrin alpha 1; spectrin alpha chain, brain; spectrin alpha chain, erythrocyte; spectrin alpha chain, erythrocytic 1; Spectrin alpha chain, non-erythrocytic 1; spectrin alpha, erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, erythrocytic 1; spectrin, alpha, erythrocytic 1 (elliptocytosis 2); spectrin, alpha, non-erythrocytic 1; spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; sph; SPH3; spherocytosis; Spna1; Spna-1; Spna2; Spna-2; SPTA; Spta1; SPTA2; SPTAN1
Gene Symbols SPTA1
Host Species Rabbit
Immunogen Recombinant protein within Human Spectrin alpha-1 aa 1-200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20739, 6708
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.